Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LMO2 Monoclonal Antibody (OTI2D7), TrueMAB™, OriGene
SDP

Mouse Monoclonal Antibody

$516.29

Specifications

Antigen LMO2
Clone OTI2D7
Concentration 1 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-167-6521
SDP
Encompass_Preferred
View Documents
OriGene
TA506177
100 μL
Each of 1 for $516.29
Only null left
Add to Cart
 
Description

Description

LMO2 Monoclonal antibody specifically detects LMO2 in Human samples. It is validated for Western Blot
Specifications

Specifications

LMO2
1 mg/mL
Monoclonal
Liquid
RUO
IgG1
-20° C, Avoid Freeze/Thaw Cycles
OTI2D7
Western Blot
Unconjugated
Mouse
Human
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.