Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LMO2 Antibody (1A9-3B11) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280835
Description
LMO2 Monoclonal specifically detects LMO2 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| LMO2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot : 0.5 ug/ml, Immunohistochemistry : 1:100-1:500, Immunocytochemistry/Immunofluorescence : 1:100-1:500, Immunoprecipitation : 1:10-1:500, Immunohistochemistry-Paraffin, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
| Cysteine-rich protein TTG-2, LIM domain only 2 (rhombotin-like 1), LIM domain only protein 2, LMO-2, RBTN2rhombotin-2, RBTNL1rhombotin-like 1, RHOM2T-cell translocation protein 2, T-cell translocation gene 2, TTG2rhombotin 2 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 1A9-3B11 | |
| Unconjugated | |
| PBS | |
| LMO2 | |
| Recombinant human LMO2. [UniProt# P25791] | |
| 0.1 mL | |
| Angiogenesis, Cancer | |
| 4005 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction