Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LMO2 Rabbit anti-Human, Unconjugated, Clone: LMO2/3147R, Abnova™

Catalog No. 89339529 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-529 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-529 Supplier Abnova Corporation Supplier No. RAB00573
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

LMO2 Monoclonal antibody specifically detects LMO2 in Human samples. It is validated for IF, Flow Cyt.

Specifications

Antigen LMO2
Applications Immunofluorescence, Flow Cytometry
Classification Monoclonal
Clone LMO2/3147R
Conjugate Unconjugated
Description LIM domain only 2 (rhombotin-like 1)
Gene LMO2
Gene Alias RBTN2, RBTNL1, RHOM2, TTG2
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 23-140 of human LMO2
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 4005
Target Species Human
Content And Storage Store at -20 to -80°C
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.