Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LMO3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598716
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598716 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598716 Supplier Invitrogen™ Supplier No. PA598716
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LMO3 Polyclonal Antibody for Western Blot, IHC (P), ELISA

The protein encoded by this gene belongs to the rhombotin family of cysteine-rich LIM domain oncogenes. This gene is predominantly expressed in the brain. Related family members, LMO1 and LMO2 on chromosome 11, have been reported to be involved in chromosomal translocations in T-cell leukemia. Many alternatively spliced transcript variants have been found for this gene. [provided by RefSeq, Aug 2011].
TRUSTED_SUSTAINABILITY

Specifications

Antigen LMO3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene LMO3
Gene Accession No. Q8TAP4
Gene Alias AI854781; BB106490; LIM domain only 3; LIM domain only 3 (rhombotin-like 2); LIM domain only protein 3; LIM only 3; Lmo3; LMO-3; Neuronal-specific transcription factor DAT1; RBTN3; Rbtn-3; RBTNL2; RGD1561357; RHOM3; Rhom-3; Rhombotin-3
Gene Symbols LMO3
Host Species Rabbit
Immunogen Recombinant Human LIM domain only protein 3 protein (1-145AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55885
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.