Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LMO3 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5143253
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
The protein encoded by this gene belongs to the rhombotin family of cysteine-rich LIM domain oncogenes. This gene is predominantly expressed in the brain. Related family members, LMO1 and LMO2 on chromosome 11, have been reported to be involved in chromosomal translocations in T-cell leukemia. Many alternatively spliced transcript variants have been found for this gene. [provided by RefSeq, Aug 2011].
Specifications
LMO3 | |
Polyclonal | |
Unconjugated | |
LMO3 | |
AI854781; BB106490; LIM domain only 3; LIM domain only 3 (rhombotin-like 2); LIM domain only protein 3; LIM only 3; Lmo3; LMO-3; Neuronal-specific transcription factor DAT1; RBTN3; Rbtn-3; RBTNL2; RGD1561357; RHOM3; Rhom-3; Rhombotin-3 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
109593, 55885 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q8BZL8, Q8TAP4 | |
LMO3 | |
Peptide with sequence C-EGLMKEGYAPQVR. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction