Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LMO3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101212
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101212 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101212 Supplier Thermo Scientific Supplier No. PA5101212
Add to Cart
Add to Cart

LMO3 Polyclonal Antibody

LMO3 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen LMO3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene LMO3
Gene Accession No. Q8BZL8, Q8TAP4, Q99MB5
Gene Alias AI854781; BB106490; LIM domain only 3; LIM domain only 3 (rhombotin-like 2); LIM domain only protein 3; LIM only 3; Lmo3; LMO-3; Neuronal-specific transcription factor DAT1; RBTN3; Rbtn-3; RBTNL2; RGD1561357; RHOM3; Rhom-3; Rhombotin-3
Gene Symbols LMO3
Host Species Rabbit
Immunogen A synthesized peptide derived from human LMO3(Accession Q8TAP4), corresponding to amino acid residues C108-R145.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109593, 497798, 55885
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.