Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LMO4 Antibody (SR1262), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322321100UL
Description
LMO4 Monoclonal antibody specifically detects LMO4 in Human, Mouse samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
LMO4 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunoprecipitation | |
SR1262 | |
Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
Breast tumor autoantigen, LIM domain only 4, LIM domain only protein 4, LIM domain transcription factor LMO4, LMO-4 | |
A synthesized peptide derived from human LMO4 (Uniprot #: P61968) | |
100 μg | |
Cancer, Tumor Suppressors | |
8543 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction