Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LMO4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986940
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6940 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6940 Supplier Bioss Supplier No. BS5966R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LMO4 encodes a cysteine-rich protein that contains two LIM domains but lacks a DNA-binding homeodomain. The encoded protein may play a role as a transcriptional regulator or as an oncogene.

Specifications

Antigen LMO4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene LMO4
Gene Accession No. P61968
Gene Alias A730077C12Rik; Breast tumor autoantigen; Crp3; ethanol induced 4; Etohi4; LIM domain only 4; LIM domain only protein 4; LIM domain transcription factor LMO4; LIM-only 4 protein; LMO4; LMO-4
Gene Symbols LMO4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human LMO4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8543
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.