Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LMO7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575815
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575815 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575815 Supplier Invitrogen™ Supplier No. PA575815
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

The LIM-only (LMO) proteins are nuclear factors characterized by a conserved LIM domain. The LIM domain contains a cysteine-rich zinc-binding motif, present in a variety of transcription factors, including the LIM homeobox (LHX) proteins expressed in the central nervous system. The deduced LMO7 protein is comprised of 1,349 amino acid residues, contains a characteristic zinc finger domain and a 3-prime UTR which possesses a short interspersed nucleotide element (SINE). RT-PCR detects predominant expression of LMO7 in heart, lung, skeletal muscle, and kidney, moderate expression in liver, ovary, brain, pancreas, and testis, and little to no expression in spleen. Research indicates that LMO7 is an afadin- and alpha-actinin-binding protein that connects the nectin-afadin and E-cadherin-catenin systems through alpha-actinin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LMO7
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene LMO7
Gene Accession No. Q8WWI1
Gene Alias C78582; F-box only protein 20; F-box protein Fbx20; FBX20; FBXO20; Gm914; KIAA0858; LIM domain 7; LIM domain only 7; LIM domain only 7 protein; LIM domain only protein 7; LMO7; LMO-7; LOMP; LOMP protein; mKIAA0858; RP11-332E3.2; TGF-beta induced protein; zinc-finger domain-containing protein
Gene Symbols LMO7
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues in Human LMO7.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361084, 380928, 4008
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.