Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LMOD1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP189396
Description
LMOD1 Polyclonal specifically detects LMOD1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
LMOD1 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
64 kDa autoantigen 1D, 64 kDa autoantigen 1D3,1D, 64 kDa autoantigen D1,64kD, D1, FLJ55689, leimodin 1 (smooth muscle), leiomodin 1 (smooth muscle), Leiomodin, muscle form, leiomodin-1, SM-Lmod, Smooth muscle leiomodin, thyroid and eye muscle autoantigen D1 (64kD), Thyroid-associated ophthalmopathy autoantigen | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
LMOD1 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:TEKQSTGVYNREAMLNFCEKETKKLMQREMSMDESKQVETKTDAKNGEERGRDASKKALGPRRDSDLGKEPK | |
0.1 mL | |
Vision | |
25802 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction