Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LMOD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555848
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555848 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555848 Supplier Invitrogen™ Supplier No. PA555848
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TEKQSTGVYN REAMLNFCEK ETKKLMQREM SMDESKQVET KTDAKNGEER GRDASKKALG PRRDSDLGKE PK Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 75%.

The leiomodin 1 protein (LMOD1) has a putative membrane-spanning region and 2 types of tandemly repeated blocks. The transcript is expressed in all tissues tested, with the highest levels in thyroid, eye muscle, skeletal muscle, and ovary. Increased expression of leiomodin 1 may be linked to Graves' disease and thyroid-associated ophthalmopathy. The leiomodin 1 protein has a putative membrane-spanning region and 2 types of tandemly repeated blocks. The transcript is expressed in all tissues tested, with the highest levels in thyroid, eye muscle, skeletal muscle, and ovary. Increased expression of leiomodin 1 may be linked to Graves' disease and thyroid-associated ophthalmopathy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LMOD1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Lmod1
Gene Accession No. P29536
Gene Alias 1D; 64 kDa autoantigen 1D; 64 kDa autoantigen 1D3; 64 kDa autoantigen D1; 64kD; 9530015K06Rik; D1; leimodin 1 (smooth muscle); leiomodin 1; leiomodin 1 (smooth muscle); leiomodin, muscle form; Leiomodin-1; LMOD1; SMLMOD; SM-Lmod; Smooth muscle leiomodin; thyroid and eye muscle autoantigen D1 (64kD); thyroid-associated ophthalmopathy autoantigen
Gene Symbols Lmod1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LMOD1. Recombinant protein control fragment (Product #RP-94429).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25802
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.