Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LMOD1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549616
Description
The leiomodin 1 protein has a putative membrane-spanning region and 2 types of tandemly repeated blocks. The transcript is expressed in all tissues tested, with the highest levels in thyroid, eye muscle, skeletal muscle, and ovary. Increased expression of leiomodin 1 may be linked to Graves′ disease and thyroid-associated ophthalmopathy. [provided by RefSeq, Jul 2008]
Specifications
LMOD1 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
P29536 | |
LMOD1 | |
KLH conjugated synthetic peptide between 214-241 amino acids from the Central region of human LMOD1. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
LMOD1 | |
Leiomodin-1, 64 kDa autoantigen 1D, 64 kDa autoantigen 1D3, 64 kDa autoantigen D1, Leiomodin, muscle form, Smooth muscle leiomodin, SM-Lmod, Thyroid-associated ophthalmopathy autoantigen, LMOD1 | |
Rabbit | |
Ammonium sulfate precipitation | |
RUO | |
25802 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction