Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LMOD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562300
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562300 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562300 Supplier Invitrogen™ Supplier No. PA562300
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RELEDIEPDR NLPVGLRQKS LTEKTPTGTF SREALMAYWE KESQKLLEKE RLGECGKVAE DKEESEEELI FTESNSEVSE EVYT Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen LMOD2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LMOD2
Gene Accession No. Q6P5Q4
Gene Alias cardiac leiomodin; CLMOD; C-Lmod; Leiomodin; leiomodin 2; leiomodin 2 (cardiac); leiomodin-2; Lmod2
Gene Symbols LMOD2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LMOD2. Recombinant protein control fragment (Product #RP-100549).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 442721
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.