Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LMOD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599989
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599989 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599989 Supplier Invitrogen™ Supplier No. PA599989
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total LMOD3.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LMOD3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene LMOD3
Gene Accession No. E9QA62, Q0VAK6
Gene Alias 5430424A14Rik; leiomodin 3; leiomodin 3 (fetal); leiomodin, fetal form; Leiomodin-3; LMOD3; NEM10; RGD1564924
Gene Symbols LMOD3
Host Species Rabbit
Immunogen A synthesized peptide derived from human LMOD3(Accession Q0VAK6), corresponding to amino acid residues P428-Y478.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 320502, 500267, 56203
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.