Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LMP7/PSMB8 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB1203329
Description
LMP7/PSMB8 Polyclonal specifically detects LMP7/PSMB8 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
LMP7/PSMB8 | |
Polyclonal | |
Unconjugated | |
PBS, with 1 mg/ml with 0.05% Sodium Azide | |
beta5i, D6S216E, EC 3.4.25.1, LMP7MGC1491, Low molecular mass protein 7, low molecular weight protein 7, Macropain subunit C13, Multicatalytic endopeptidase complex subunit C13, protease component C13, proteasome (prosome, macropain) subunit, beta type, 8 (large multifunctionalpeptidase 7), proteasome catalytic subunit 3i, Proteasome component C13, proteasome subunit beta 5i, proteasome subunit beta type-8, Proteasome subunit beta-5i, proteasome subunit Y2, proteasome-related gene 7, PSMB5iD6S216, Really interesting new gene 10 protein, RING10proteasome (prosome, macropain) subunit, beta type, 8 (large multifunctionalprotease 7), Y2 | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human and Mouse. Predicted cross-reactivity bsaed on sequence identity: Rat (100%) and Sheep (100%). | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence | |
1 mg/mL | |
Western Blot 1.0 ug/ml, Immunocytochemistry/Immunofluorescence 5 ug/ml | |
P28062 | |
PSMB8 | |
Synthetic Peptide: C V(259) E S T D V S D L L H Q Y R E A(274) | |
100 μg | |
Stem Cell Markers | |
5696 | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction