Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LMTK2 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP232182F
Description
LMTK2 Polyclonal antibody specifically detects LMTK2 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
LMTK2 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
AATYK2FLJ46659, Apoptosis-associated tyrosine kinase 2, Brain-enriched kinase, BREKCDK5/p35-regulated kinase, cprk, EC 2.7.11, KIAA1079hBREK, kinase phosphatase inhibitor 2, Kinase/phosphatase/inhibitor 2, KPI-2, KPI2EC 2.7.11.1, lemur tyrosine kinase 2Serine/threonine-protein kinase KPI-2, LMR2cyclin-dependent kinase 5/p35-regulated kinase, serine/threonine-protein kinase LMTK2 | |
The immunogen this antibody was made to, maps to a region between residue 675 to 725 of human Lemur Tyrosine Kinase 2 using the numbering given in entry NP_055731.2 | |
0.1 mL | |
Protein Kinase | |
22853 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
FITC | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction