Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LMTK2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106513
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106513 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106513 Supplier Invitrogen™ Supplier No. PA5106513
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total LMTK2.

Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. The tyrosine kinase (TK) group is mainly involved in the regulation of cell-cell interactions such as differentiation, adhesion, motility and death. There are currently about 90 TK genes sequenced, 58 are of receptor protein TK (e.g. EGFR, EPH, FGFR, PDGFR, TRK, and VEGFR families), and 32 of cytosolic TK (e.g. ABL, FAK, JAK, and SRC families).
TRUSTED_SUSTAINABILITY

Specifications

Antigen LMTK2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene LMTK2
Gene Accession No. Q3TYD6, Q8IWU2
Gene Alias 2900041G10Rik; A330101P12Rik; aatyk2; Apoptosis-associated tyrosine kinase 2; brain-enriched kinase; BREK; CDK5/p35-regulated kinase; cprk; cyclin-dependent kinase 5/p35-regulated kinase; FLJ46659; hBREK; Kiaa1079; kinase phosphatase inhibitor 2; kinase/phosphatase/inhibitor 2; KPI2; KPI-2; KPI-2 protein; Lemur tyrosine kinase 2; LMKT2; LMR2; Lmtk2; PPP1R100; protein phosphatase 1, regulatory subunit 100; serine/threonine-protein kinase KPI-2; Serine/threonine-protein kinase LMTK2
Gene Symbols LMTK2
Host Species Rabbit
Immunogen A synthesized peptide derived from human LMTK2(Accession Q8IWU2), corresponding to amino acid residues K665-N715.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22853, 231876
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.