Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LMX1A Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP2411930.025MG
Description
LMX1A Polyclonal specifically detects LMX1A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
LMX1A | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
LMX1A | |
Antibody was raised against a 16 amino acid peptide near the carboxy terminus of human LMX1A. The immunogen is located within amino acids 300 - 350 of LMX1A. Amino Acid Squence: CPQMPGDHMHPYGAEPL | |
Affinity Purified | |
RUO | |
4009 | |
Human, Mouse, Rat | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1 - 2 μg/mL, ELISA, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL | |
LIM homeobox transcription factor 1, alpha, LMX1.1 | |
Rabbit | |
42 kDa | |
0.025 mg | |
Primary | |
At least two isoforms are known to exist. This antibody is predicted to not cross-react with LMX1B. | |
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction