Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LMX1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567308
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567308 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567308 Supplier Invitrogen™ Supplier No. PA567308
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DIATGPESLER CFPRGQTDCA KMLDGIKMEE HALRPGPATL GVLLGSDCPH PAVCEGCQRP ISDRFL Highest antigen sequence identity to the following orthologs - mouse 67%, rat 100%.

LMX1B, LIM homeobox transcription factor 1B, belongs to the LIM-homeodomain family. Members of this family are known to be important for pattern formation during development. LMX1B regulates mid-hindbrain patterning LMX1B-null mice embryos have a severe reduction in the number of midbrain dopaminergic neurons compared to wild-type. While LMX1B appears to be important for both the development and the survival of dopamine neurons, the related LMX1A is crucial for the differentiation of these cells. However, LMX1A and LMX1B function cooperatively to regulated the proliferation, specification and differentiation of midbrain dopaminergic neuronal progenitors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LMX1B
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Lmx1b
Gene Accession No. O60663
Gene Alias LIM homeo box transcription factor 1B; LIM homeobox transcription factor 1 beta; LIM homeobox transcription factor 1, beta; LIM homeobox transcription factor 1-beta; LIM/homeobox protein 1.2; LIM/homeobox protein LMX1B; LMX1.1; LMX1.2; LMX-1.2; LMX1B; NPS1
Gene Symbols Lmx1b
Host Species Rabbit
Immunogen Recombinant Human LMX1B. Recombinant protein control fragment (Product #RP-107946).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4010
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.