Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LMX1B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578395
Description
Positive Control: mouse skeletal muscle Predicted Reactivity: Human (100%), Pig (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
LMX1B, LIM homeobox transcription factor 1B, belongs to the LIM-homeodomain family. Members of this family are known to be important for pattern formation during development. LMX1B regulates mid-hindbrain patterning LMX1B-null mice embryos have a severe reduction in the number of midbrain dopaminergic neurons compared to wild-type. While LMX1B appears to be important for both the development and the survival of dopamine neurons, the related LMX1A is crucial for the differentiation of these cells. However, LMX1A and LMX1B function cooperatively to regulated the proliferation, specification and differentiation of midbrain dopaminergic neuronal progenitors.
Specifications
LMX1B | |
Polyclonal | |
Unconjugated | |
Lmx1b | |
LIM homeo box transcription factor 1B; LIM homeobox transcription factor 1 beta; LIM homeobox transcription factor 1, beta; LIM homeobox transcription factor 1-beta; LIM/homeobox protein 1.2; LIM/homeobox protein LMX1B; LMX1.1; LMX1.2; LMX-1.2; LMX1B; NPS1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
114501, 16917, 4010 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Frozen), Western Blot | |
0.2 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
O60663, O88609 | |
Lmx1b | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human LMX1B. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction