Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LNK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552154
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552154 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552154 Supplier Invitrogen™ Supplier No. PA552154
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FDPPKSSRPK LQAACSSIQE VRRCTRLEMP DNLYTFVLKV KDRTDIIFEV GDEQQLNSWM AELSECTGRG LESTEAEMHI PSALEPSTSS SPRGSTDSLN QGASPGGLLD PACQKTDHFL SCYPWFH Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 77%.

This gene encodes a member of the SH2B adaptor family of proteins, which are involved in a range of signaling activities by growth factor and cytokine receptors. The encoded protein is a key negative regulator of cytokine signaling and plays a critical role in hematopoiesis. Mutations in this gene have been associated with susceptibility to celiac disease type 13 and susceptibility to insulin-dependent diabetes mellitus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LNK
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SH2B3
Gene Accession No. Q9UQQ2
Gene Alias AI429800; IDDM20; linker of T-cell receptor pathways; LNK; Lymphocyte adapter protein; lymphocyte-specific adapter protein Lnk; SH2B adapter protein 3; SH2B adaptor protein 3; Sh2b3; Signal transduction protein Lnk
Gene Symbols SH2B3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LNK. Recombinant protein control fragment (Product #RP-102117).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10019
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.