Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LNK/SH2B3 Antibody (OTI2D8), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272514MFV450
Description
LNK/SH2B3 Monoclonal antibody specifically detects LNK/SH2B3 in Human samples. It is validated for Flow CytometrySpecifications
LNK/SH2B3 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
10019 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry | |
OTI2D8 | |
Flow Cytometry | |
IDDM20, LNKSH2B adapter protein 3, Lymphocyte adapter protein, Lymphocyte-specific adapter protein Lnk, SH2B adaptor protein 3, Signal transduction protein Lnk | |
Full length human recombinant protein of human SH2B3 (NP_005466) produced in HEK293T cell. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction