Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LNPEP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583672
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583672 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583672 Supplier Invitrogen™ Supplier No. PA583672
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EALIHQLKIN PYVLSDKDRA NLINNIFELA GLGKVPLKRA FDLINYLGNE NHTAPITEAL FQTDLIYNLL EKLGYMDLAS RLVTRVFKLL QNQIQQQTWT DEGTPSMREL RSALLEFACT HNLGNCSTTA MKL Highest antigen sequence indentity to the following orthologs: Mouse - 85%, Rat - 87%.

This gene encodes a zinc-dependent aminopeptidase that cleaves vasopressin, oxytocin, lys-bradykinin, met-enkephalin, dynorphin A and other peptide hormones. The protein can be secreted in maternal serum, reside in intracellular vesicles with the insulin-responsive glucose transporter GLUT4, or form a type II integral membrane glycoprotein. The protein catalyzes the final step in the conversion of angiotensinogen to angiotensin IV (AT4) and is also a receptor for AT4. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LNPEP
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LNPEP
Gene Accession No. Q9UIQ6
Gene Alias 2010309L07Rik; 4732490P18Rik; aminopeptidase Vp165; angiotensin IV receptor; AT (4) receptor; CAP; Cystinyl aminopeptidase; GP160; insulin-regulated aminopeptidase; insulin-regulated membrane aminopeptidase; insulin-regulated membrane aminopeptidase IRAP; insulin-responsive aminopeptidase; IRAP; leucyl and cystinyl aminopeptidase; leucyl/cystinyl aminopeptidase; leucyl-cystinyl aminopeptidase; Leucyl-cystinyl aminopeptidase, pregnancy serum form; LNPEP; OTase; oxytocinase; Placental leucine aminopeptidase; PLAP; P-LAP; vasopressinase; vesicle protein of 165 kDa; vp160; vp165
Gene Symbols LNPEP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LNPEP. Recombinant protein control fragment (Product #RP-90122).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4012
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.