Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LNX1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542348
Description
Peptide sequence: SHREWDLPIY VISVEPGGVI SRDGRIKTGD ILLNVDGVEL TEVSRSEAVA Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.
Lnx1 (Ligand of Numb Protein X 1) protein is encoded by the Lnx1 gene located on chromosome 4 in humans and is known as PDZRN2 (PDZ domain-containing RING finger 2). It is through its RING finger and PDZ domains that Lnx1 forms oligomers and interacts with some of its target proteins such as JAM4 to form multiprotein complexes. Lnx1 is best studied as a E3 ubiquitin ligase that also contains the NPXY motifs which allows binding to the Phosphotyrosine binding domain (PTB) on Numb protein. This interaction has been shown to be responsible for ubiquitination of Numb, which prevents the proteasomal degradation of Notch intracellular domain (NICD) and thus positively regulates the Notch signaling pathway.
Specifications
LNX1 | |
Polyclonal | |
Unconjugated | |
LNX1 | |
E3 ubiquitin-protein ligase LNX; Ligand of Numb protein X 1; ligand of Numb-binding protein 1; Ligand of Numb-protein X 1; ligand of numb-protein X 1, E3 ubiquitin protein ligase; LNX; Lnx1; MPDZ; multi-PDZ-domain-containing protein, E3 ubiquitin-protein ligase LNX; numb-binding protein 1; PDZ domain-containing RING finger protein 2; PDZRN2; RING-type E3 ubiquitin transferase LNX; UNQ574/PRO1136 | |
Rabbit | |
Protein A | |
RUO | |
84708 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8TBB1 | |
LNX1 | |
Synthetic peptide directed towards the C-terminal of human LNX1 (aa 527-576). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction