Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LNX2 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298102APC
Description
LNX2 Polyclonal antibody specifically detects LNX2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
LNX2 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
FLJ23932, FLJ38000, ligand of Numb protein X 2, ligand of numb-protein X 2, MGC46315, Numb-binding protein 2, PDZ domain containing ring finger 1, PDZ domain-containing RING finger protein 1, PDZRN1FLJ12933 | |
Produced in rabbits immunized with E. coli-derived Human LNX2 fragment. | |
0.1 mL | |
Growth and Development, Neuronal Cell Markers, Zinc Finger | |
222484 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction