Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LNX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546740
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546740 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546740 Supplier Invitrogen™ Supplier No. PA546740
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LRERRFGNRA HNHSDSNSPR EEIFQVALHK RDSGEQLGIK LVRRTDEPGV.

LNX2 (Ligand of Numb Protein X 2) protein is encoded by the LNX2 gene located on chromosome 13 in humans and is known as PDZRN1 (PDZ domain-containing RING finger 1). It is through its PDZ domains that LNX2 forms oligomers and interacts with some of its target proteins. LNX2 is best studied as a E3 ubiquitin ligase that also contains the NPXY motifs which allows binding to the Numb protein. This interaction has been shown to be responsible for ubiquitination of Numb, which prevents the proteasomal degradation of Notch intracellular domain (NICD) and thus modulates the Notch signaling pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LNX2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LNX2
Gene Accession No. Q8N448
Gene Alias 9630046H24; AW209022; ligand of Numb protein X 2; ligand of numb-protein X 2; Lnx2; Numb-binding protein 2; PDZ domain containing ring finger 1; PDZ domain-containing RING finger protein 1; PDZRN1
Gene Symbols LNX2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human LNX2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 222484
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.