Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LNX2 Polyclonal Antibody

Catalog No. PI720004
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI720004 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI720004 Supplier Invitrogen™ Supplier No. 720004

Rabbit Polyclonal Antibody

This antibody is predicted to react with Goat, Bovine, Horse, Pig, Cat and Rabbit.

LNX2 (Ligand of Numb Protein X 2) protein is encoded by the LNX2 gene located on chromosome 13 in humans and is known as PDZRN1 (PDZ domain-containing RING finger 1). It is through its PDZ domains that LNX2 forms oligomers and interacts with some of its target proteins. LNX2 is best studied as a E3 ubiquitin ligase that also contains the NPXY motifs which allows binding to the Numb protein. This interaction has been shown to be responsible for ubiquitination of Numb, which prevents the proteasomal degradation of Notch intracellular domain (NICD) and thus modulates the Notch signaling pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LNX2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.2
Gene LNX2
Gene Accession No. Q8N448, Q91XL2
Gene Alias 9630046H24; AW209022; ligand of Numb protein X 2; ligand of numb-protein X 2; Lnx2; Numb-binding protein 2; PDZ domain containing ring finger 1; PDZ domain-containing RING finger protein 1; PDZRN1
Gene Symbols LNX2
Host Species Rabbit
Immunogen Multiple peptides against Human LNX2 (aa 442-458, 129-145, 671-683).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140887, 222484, 360761
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.