Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LOC113509844 Polyclonal Antibody, Invitrogen™

Catalog No. FEPA5144399
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144399 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144399 Supplier Thermo Scientific Supplier No. PA5144399

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

LOC113509844 is a gene that is currently annotated as a long non-coding RNA (lncRNA) in the human genome. Long non-coding RNAs are RNA molecules that are longer than 200 nucleotides and do not encode proteins. They have been found to play important roles in various cellular processes, including gene regulation, chromatin remodeling, and cellular development. The specific function of LOC113509844 is not yet fully understood, and further research is required to elucidate its role in cellular processes and its potential implications for human health and disease. It is worth noting that lncRNAs have been implicated in various biological processes and diseases, including cancer, cardiovascular diseases, and neurological disorders. Therefore, understanding the specific functions and mechanisms of lncRNAs like LOC113509844 may provide valuable insights into the molecular mechanisms underlying these conditions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LOC113509844
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.735 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene Alias uncharacterized protein LOC113509844
Host Species Rabbit
Immunogen Recombinant Galleria mellonella LOC113509844 protein (39-230aa )
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Insect
Content And Storage -20°C or -80°C if preferred
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.