Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LOC113516003 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5144400

Description
LOC113516003 is a gene that is part of the human genome. It is an example of a gene identifier or accession number, which is used to uniquely identify specific genes in genetic databases. However, without further context or information, it is difficult to provide specific details about the function or characteristics of LOC113516003.
Specifications
LOC113516003 | |
Polyclonal | |
Unconjugated | |
LOC113516003, serine protease inhibitor dipetalogastin-like | |
Recombinant Galleria mellonella LOC113516003 protein (19-219aa ) | |
100 μL | |
Primary | |
-20°C or -80°C if preferred | |
IgG |
ELISA, Western Blot | |
3.958 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
Rabbit | |
Protein G | |
RUO | |
Insect | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction