Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LOC281370, Mouse anti-Bovine, R-PE, Clone: 5B9-B3, Abnova™

Catalog No. 89338030 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-338-030 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-338-030 Supplier Abnova Corporation Supplier No. MAB18105
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifications

Antigen LOC281370
Applications ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 5B9-B3
Conjugate R-PE
Dilution ELISA Immunocytochemistry (1:100) Immunofluorescence (1:100) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide).
Gene polyubiquitin
Gene Alias -
Gene Symbols LOC281370
Host Species Mouse
Immunogen Native bovine ubiquitin, conjugated to KLH.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 281370
Target Species Bovine, Human
Content And Storage Store at -20°C.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.