Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAF11L2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571796
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571796 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571796 Supplier Invitrogen™ Supplier No. PA571796
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TAF11L2 Polyclonal Antibody for Western Blot, Flow

Predicted to enable transcription coactivator activity. Predicted to be involved in RNA polymerase II preinitiation complex assembly. Predicted to be part of transcription factor TFIID complex. [provided by Alliance of Genome Resources, Apr 2022].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAF11L2
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 1.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene LOC391742
Gene Accession No. 0
Gene Alias LOC391742; YE016
Gene Symbols LOC391742
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 59-87 amino acids from the Central region of human YE016.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 391742
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.