Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LOC4327476 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109331
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109331 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109331 Supplier Invitrogen™ Supplier No. PA5109331
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: Rice stem.

Has oxidoreductase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LOC4327476
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4
Gene LOC4327476
Gene Accession No. 0
Gene Alias aldo-keto reductase family 4 member C10; LOC4327476; Os01g0847700; OSNPB_010847700; P0005H10.17; putative aldose reductase
Gene Symbols LOC4327476
Host Species Rabbit
Immunogen Synthetic peptide within Rice Os01g0847700 aa 262-311.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4327476
Target Species Plant
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.