Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LOC81691 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559283
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559283 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559283 Supplier Invitrogen™ Supplier No. PA559283
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KLKFLAKVIL GKDIQCPDRL GHDATEDART ILELARYFLK HGPKKIAELN LEALANHQEI QAAGQEPKNT AEVLQHPNTS VLECLDSVGQ KLLFLTRETD AGELPSSRNC QTIKCLS Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 67%.

REXO5 (RNA Exonuclease 5) is a Protein Coding gene. Among its related pathways are Ribosome biogenesis in eukaryotes. An important paralog of this gene is REXO1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LOC81691
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene REXO5
Gene Accession No. Q96IC2
Gene Alias 44M2.3; Exonuclease NEF-sp; LOC81691; putative RNA exonuclease NEF-sp; REXO5; RNA exonuclease 5
Gene Symbols REXO5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LOC81691.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81691
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.