Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BINDER™ Locking Latch for Access Port Plug
SDP

Catalog No. 08112030 Shop All BINDER Inc. Products
Click to view available options
For Use With (Equipment):
BINDER Chambers Series LIT MK

This Locking Latch for Access Port Plug is an option for BINDER™ LIT MK Series.

  • 30 mm diameter
  • For securing silicone plug of an access port
  • Maximum Number of Shelves: 11

Specifications

Description Locking latch for access port plug - 30 mm diameter, For securing the silicone plug of an access port
For Use With (Equipment) BINDER Chambers Series LIT MK
Material Steel
No. of Shelves 1
Type Locking Latch
Diameter (English) 1.2 in.
Diameter (Metric) 30 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.