Learn More
Post Medical Locking-Top Sharps Containers
Catalog No. p-3238071
Click to view available options
Capacity (English):
1.5 qt.
2 qt.
1.5 gal.
2.5 gal.
Catalog No. PMSL200
Supplier: Post Medical SL200
Only null left
Add to Cart
Add to Cart
Safe, one-handed sharps disposal
For use in patient rooms and all hospital departments including laboratory.
  • Thick-walled, puncture-resistant sharps containment systems provide safe, convenient and affordable site of origin sharps disposal
  • Vertical drop disposal minimizes excessive handling; reduces risks associated with horizontal “mail-box” type disposal
  • Once self-contained screw cap is secured and locked, leakage of fluids during handling, transport and disposal is virtually eliminated
  • 1.0, 1.5 and 2.5 gal. Containers
  • Can be used freestanding or with metal stands or cabinets
  • Bulk 3.5 and 6 gal. Containers
  • Designed for freestanding use only
Compliance
Meets OSHA requirements.
Specifications
Capacity (Metric) 1.9 L
Closure Type Leaktight Locking Top, Snap-Tite Lid, Safe-T-Ring
Capacity (English) 2 qt.
For Use With (Application) For use in patient rooms and all hospital departments including laboratory.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.