Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermco™ LogTag™ Dry Ice Probe-less Low Temperature Recorder

Catalog No. 04500007
Change view
Click to view available options
Accuracy (Temperature):
0.5°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Accuracy (Temperature)
04-500-007 0.5°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-500-007 Supplier Thermco™ Supplier No. LTSRIL8
Add to Cart
Add to Cart

Record temperatures in harsh enviroments, such as dry ice. Thermco™ Products LogTag™ Dry Ice Probe-less Low Temperature Recorder measures and stores up to 8000temperature readings in temperature enviroments down to -80°C.

  • N.I.S.T. Compliant
  • Probe-less temperature recordrer
  • Push Button Logging start with optional delayed start or specific time and date start
  • Single use recorder high performance at low cost
  • Alert indicator indicates

Certifications

N.I.S.T. Compliant

Warranty and Services

1 Year

Specifications

Type Recorder, Dry Ice Probe-less Low Temperature
Description Single use data logger
Requirements Needs LTIUSB Docking station to download data
For Use With (Application) Record temperatures in shipping containers utilizing Dry Ice
Accuracy (Temperature) 0.5°C
Dimensions (L x W x H) 86 x 54 x 8.6 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.