Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LOH12CR1 Antibody (OTI1E1), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272518G
Description
LOH12CR1 Monoclonal specifically detects LOH12CR1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
LOH12CR1 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
BORCS5 | |
Full length recombinant protein of human LOH12CR1 (NP_477517) produced in HEK293T cells. | |
0.1 mL | |
118426.0 | |
Store at 4°C in the dark. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI1E1 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
LOH1CR12, loss of heterozygosity 12 chromosomal region 1 protein, loss of heterozygosity, 12, chromosomal region 1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction