Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LOH12CR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100604
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100604 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100604 Supplier Invitrogen™ Supplier No. PA5100604
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total LOH12CR1.

LOH12CR1 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LOH12CR1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene BORCS5
Gene Accession No. Q969J3, Q9D920
Gene Alias 5830457J20Rik; AW124643; BLOC-1 related complex subunit 5; BLOC-1-related complex subunit 5; BORCS5; Loh12cr1; LOH1CR12; Loss of heterozygosity 12 chromosomal region 1; loss of heterozygosity 12 chromosomal region 1 protein; loss of heterozygosity 12 chromosomal region 1 protein homolog; loss of heterozygosity, 12, chromosomal region 1; loss of heterozygosity, 12, chromosomal region 1 homolog (human); Myristoylated lysosomal protein; Myrlysin
Gene Symbols BORCS5
Host Species Rabbit
Immunogen A synthesized peptide derived from human BORCS5(Accession Q969J3), corresponding to amino acid residues P22-G72.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 118426, 362452, 67774
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.