Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LOH12CR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558821
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558821 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558821 Supplier Invitrogen™ Supplier No. PA558821
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSVETLFSFM QERQKRYAKY AEQIQKVNEM SAILRRIQMG IDQTVPLLDR LNSMLPEGER LEPFSMKPDR ELRL Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 88%.

LOH12CR1 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LOH12CR1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BORCS5
Gene Accession No. Q969J3
Gene Alias 5830457J20Rik; AW124643; BLOC-1 related complex subunit 5; BLOC-1-related complex subunit 5; BORCS5; Loh12cr1; LOH1CR12; Loss of heterozygosity 12 chromosomal region 1; loss of heterozygosity 12 chromosomal region 1 protein; loss of heterozygosity 12 chromosomal region 1 protein homolog; loss of heterozygosity, 12, chromosomal region 1; loss of heterozygosity, 12, chromosomal region 1 homolog (human); Myristoylated lysosomal protein; Myrlysin
Gene Symbols BORCS5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LOH12CR1. Recombinant protein control fragment (Product #RP-97841).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 118426
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.