Learn More
Invitrogen™ LOH12CR1 Polyclonal Antibody

Description
Immunogen sequence: VIKLQEIPTF QPLLKGLLSG QTSPTNAKLE KLDSQQVLQL CLRYQDHLHQ CAEAVAFDQN ALVKRIKEMD L Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 99%.
Specifications
Specifications
| Antigen | LOH12CR1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | BORCS5 |
| Gene Accession No. | Q969J3 |
| Gene Alias | 5830457J20Rik; AW124643; BLOC-1 related complex subunit 5; BLOC-1-related complex subunit 5; BORCS5; Loh12cr1; LOH1CR12; Loss of heterozygosity 12 chromosomal region 1; loss of heterozygosity 12 chromosomal region 1 protein; loss of heterozygosity 12 chromosomal region 1 protein homolog; loss of heterozygosity, 12, chromosomal region 1; loss of heterozygosity, 12, chromosomal region 1 homolog (human); Myristoylated lysosomal protein; Myrlysin |
| Gene Symbols | BORCS5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.