Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Lomeguatrib
GREENER_CHOICESDP

Catalog No. 435910 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-591-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 43-591-0 Supplier R&D Systems™ Supplier No. 4359/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. MGMT inhibitor

Lomeguatrib (LomG) is a potent inhibitor of O6-methylguanine-DNA methyltransferase (MGMT); (IC50 = 0.009 μM in cell-free extracts from HeLa S3 cells); LomG attenuates MGMT activity in vitro and in vivo. LomG enhances the antitumor activity of temozolomide (Cat. No. 2706) in both human melanoma and MCF-7 xenografts. LomG acts as a pseudosubstrate for MGMT and can be used to induce rapid ubiquitin proteasome pathway degradation of proteins that have a MGMT fusion.

Chemical Identifiers

CAS 192441-08-0
Molecular Formula C10H8BrN5OS

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 326.17
Purity >98%
Molecular Weight (g/mol) 326.17
Chemical Name or Material Lomeguatrib
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.