Learn More
Invitrogen™ Loncastuximab Chimeric Recombinant Human Monoclonal Antibody

Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Specifications
Specifications
| Antigen | Loncastuximab Chimeric |
| Applications | ELISA |
| Classification | Recombinant Monoclonal |
| Concentration | 4.55 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.94mM citric acid, 16.7mM sodium citrate with 6.2% sucrose, 0.01% Tween 80 and no preservative; pH 6.5 |
| Gene Alias | RB4v1.2 |
| Host Species | Human |
| Immunogen | Human CD19. |
| Purification Method | Protein A |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.