Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LONP1 Antibody (LON20/H1), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP20478A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP20478A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP20478A70 Supplier Novus Biologicals Supplier No. NBP250478AF750
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LONP1 Monoclonal specifically detects LONP1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Knockdown Validated.

Specifications

Antigen LONP1
Applications Western Blot, ELISA, Immunohistochemistry, KnockDown
Classification Monoclonal
Clone LON20/H1
Conjugate Alexa Fluor 750
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias EC 3.4.21, EC 3.4.21.-, EC 3.4.21.53, hLON, hLON ATP-dependent protease, LON, lon peptidase 1, mitochondrial, lon protease homolog, mitochondrial, Lon protease-like protein, LONHs, LONP, Mitochondrial ATP-dependent protease Lon, mitochondrial lon peptidase 1, mitochondrial lon protease-like protein, PIM1, protease, serine, 15, PRSS15MGC1498, Serine protease 15
Gene Symbols LONP1
Host Species Mouse
Immunogen KLH-peptide conjugate - N-CEKDDKDAIEEKFRERLKE-C
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cancer, Epigenetics, metabolism
Primary or Secondary Primary
Gene ID (Entrez) 9361
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.