Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LONP1 Antibody (LON20/H1), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250478C
Description
LONP1 Monoclonal specifically detects LONP1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Knockdown Validated.Specifications
LONP1 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
LONP1 | |
KLH-peptide conjugate - N-CEKDDKDAIEEKFRERLKE-C | |
0.1 mL | |
Cancer, Epigenetics, metabolism | |
9361 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunohistochemistry, KnockDown | |
LON20/H1 | |
Western Blot, ELISA, Immunohistochemistry | |
EC 3.4.21, EC 3.4.21.-, EC 3.4.21.53, hLON, hLON ATP-dependent protease, LON, lon peptidase 1, mitochondrial, lon protease homolog, mitochondrial, Lon protease-like protein, LONHs, LONP, Mitochondrial ATP-dependent protease Lon, mitochondrial lon peptidase 1, mitochondrial lon protease-like protein, PIM1, protease, serine, 15, PRSS15MGC1498, Serine protease 15 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction