Learn More
Invitrogen™ LONP2 Polyclonal Antibody

Description
Recommended positive controls: mouse brain. Predicted reactivity: Mouse (96%), Zebrafish (87%), Xenopus laevis (92%), Chicken (93%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | LONP2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | LONP2 |
| Gene Accession No. | Q86WA8, Q9DBN5 |
| Gene Alias | 1300002A08Rik; AU015403; HAMAP-Rule:MF_03121}; Lon peptidase 2; lon peptidase 2, peroxisomal; lon protease 2; lon protease 2 {ECO:0000255; Lon protease homolog 2, peroxisomal; lon protease homolog 2, peroxisomal {ECO:0000255; lon protease-like protein 2; lon protease-like protein 2 {ECO:0000255; LONP; Lonp2; LONPL; LRRGT00016; MGC4840; Peroxisomal Lon protease; peroxisomal Lon protease {ECO:0000255; peroxisomal Lon protease homolog 2; peroxisomal LON protease like; Peroxisomal Lon protease-like protein 2-like protein; pLon; RGD1305466 |
| Gene Symbols | LONP2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.