Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LONP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109922
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109922 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109922 Supplier Invitrogen™ Supplier No. PA5109922
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSSVSPIQIP SRLPLLLTHE GVLLPGSTMR TSVDSARNLQ LVRSRLLKGT SLQSTILGVI PNTPDPASDA QDLPPLHRIG TAALAVQVVG SNWPKPHYTL LITGLCRFQI VQVLKEKPYP IAEVEQLDRL EEFPNTCKMR EELGELSEQF YKYAVQLVEM LDMSVPAVAK LRRLLDSLPR EALPDILTSI IRTSNKEKLQ ILDAVSLEER FKMTIPLLVR QIEGLKLLQK TRKPKQDDDK RVIAIRPIRR ITHISGTLED EDEDEDNDDI VMLEKKIRTS SMPEQAHKVC VKEIKRLKKM.

ATP-dependent serine protease that mediates the selective degradation of misfolded and unassembled polypeptides in the peroxisomal matrix. Necessary for type 2 peroxisome targeting signal (PTS2)-containing protein processing and facilitates peroxisome matrix protein import.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LONP2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.93 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene LONP2
Gene Accession No. Q3MIB4, Q86WA8, Q9DBN5
Gene Alias 1300002A08Rik; AU015403; HAMAP-Rule:MF_03121}; Lon peptidase 2; lon peptidase 2, peroxisomal; lon protease 2; lon protease 2 {ECO:0000255; Lon protease homolog 2, peroxisomal; lon protease homolog 2, peroxisomal {ECO:0000255; lon protease-like protein 2; lon protease-like protein 2 {ECO:0000255; LONP; Lonp2; LONPL; LRRGT00016; MGC4840; Peroxisomal Lon protease; peroxisomal Lon protease {ECO:0000255; peroxisomal Lon protease homolog 2; peroxisomal LON protease like; Peroxisomal Lon protease-like protein 2-like protein; pLon; RGD1305466
Gene Symbols LONP2
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 291922, 66887, 83752
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.