Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LONRF1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | LONRF1 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
Description
LONRF1 Polyclonal specifically detects LONRF1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| LONRF1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| LON peptidase N-terminal domain and ring finger 1, LON peptidase N-terminal domain and RING finger protein 1, RING finger protein 191, RNF191FLJ23749 | |
| LONRF1 | |
| IgG | |
| Protein A purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Zinc Finger | |
| 91694 | |
| Synthetic peptides corresponding to LONRF1(LON peptidase N-terminal domain and ring finger 1) The peptide sequence was selected from the N terminal of LONRF1. Peptide sequence MSSPAVARTSPGGSREMAPAPQGRGRFWEVGGGSGHRLERAAAESERWEL. | |
| Primary | |
| 46 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title