Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LONRF1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554831
Description
Immunogen sequence: MEESQSLNEP SPKQSEEIPE VTSEPVKGSL NRAQSAQSIN STEMPAREDC LKRVSSEPVL SVQE Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 63%.
The function remains unknown.
Specifications
LONRF1 | |
Polyclonal | |
Unconjugated | |
LONRF1 | |
LON peptidase N-terminal domain and ring finger 1; LON peptidase N-terminal domain and RING finger protein 1; LONRF1; RGD1562583; RING finger protein 191; RNF191 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
91694 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q17RB8 | |
LONRF1 | |
Recombinant protein corresponding to Human LONRF1. Recombinant protein control fragment (Product #RP-93828). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction