Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LONRF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542356
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542356 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542356 Supplier Invitrogen™ Supplier No. PA542356
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MSSPAVARTS PGGSREMAPA PQGRGRFWEV GGGSGHRLER AAAESERWEL Sequence homology: Human: 100%; Mouse: 86%; Rat: 86%.

The function remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LONRF1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LONRF1
Gene Accession No. Q17RB8
Gene Alias LON peptidase N-terminal domain and ring finger 1; LON peptidase N-terminal domain and RING finger protein 1; LONRF1; RGD1562583; RING finger protein 191; RNF191
Gene Symbols LONRF1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human LONRF1 (aa 1-50).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91694
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.