Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LONRF2 Monoclonal Antibody (1C7)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA523142
Description
Peptide Sequence: MKNYEQALQD ASAACQNEPL LIKGHQVKAQ ALSGLGRSKE VLKEFLYCLA LNPECNSVKK EAQKVMCEVL FSATANVHEN LTSSIQSRLK AQGHSHMNAQ ALLEEGDA.
LONRF2 contains one Lon domain, one RING type zinc finger and six TPR repeats. The function of LONRF2 is unknown. There are two named isoforms.
Specifications
LONRF2 | |
Monoclonal | |
0.2-1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q1L5Z9 | |
LONRF2 | |
LONRF2 (NP_940863, 1 a.a. ~ 108 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA | |
1C7 | |
Unconjugated | |
LONRF2 | |
LON peptidase N-terminal domain and ring finger 2; LON peptidase N-terminal domain and RING finger protein 2; LONRF2; Neuroblastoma apoptosis-related protease; RING finger protein 192; RNF192 | |
Mouse | |
Affinity Chromatography | |
RUO | |
164832 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction